EMAIL THIS PAGE TO A FRIEND
Drug and chemical toxicology
Investigation on acute toxicity and behavioral changes in Channa punctatus (Bloch) due to organophosphate pesticide profenofos.
PMID 21770742
Abstract
Acute toxicity of an organophosphate pesticide profenofos (O-4-bromo-2- chlorophenyl-O-ethyl S-propyl phosphorothioate) to freshwater fish, Channa punctatus (Bloch), was studied in a static bioassay. Estimated 96-hour LC(50) of profenofos was found to be 2.68 μgL(-1). On the basis of the obtained LC(50) values for 96-hour exposure intervals, profenofos can be rated as highly toxic to C. punctatus. Fish exposed to profenofos showed hyper excitability, discoloration, erratic swimming, and secretion of excess amounts of mucus on the body and gills with eventual exhaustion and death.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45632
Profenofos, PESTANAL®, analytical standard
C11H15BrClO3PS
PS1024
Profenofos, analytical standard
C11H15BrClO3PS