EMAIL THIS PAGE TO A FRIEND
Mycoses
Case report: onychomycosis due to Trichophyton schoenleinii.
PMID 21771109
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0415
Ciclopirox olamine, analytical standard
C12H17NO2 · C2H7NO
C2162700
Ciclopirox olamine, European Pharmacopoeia (EP) Reference Standard
C12H17NO2 · C2H7NO