EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Observations on the remarkable (and mysterious) wound-healing process of the bottlenose dolphin.
PMID 21776005
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W310204
Isovaleric acid, ≥99%, FCC, FG
C5H10O2
W310212
Isovaleric acid, natural, ≥98%, FG
C5H10O2