EMAIL THIS PAGE TO A FRIEND
Veterinary parasitology
Efficacy of a novel topical combination of fipronil, amitraz and (S)-methoprene for treatment and control of induced infestations of brown dog ticks (Rhipicephalus sanguineus) on dogs.
PMID 21777733
Abstract
Four laboratory studies were conducted to demonstrate that a single topical dose of a novel spot-on combination containing fipronil, amitraz and (S)-methoprene (CERTIFECT™, Merial Limited, GA, USA) is efficacious against the brown dog tick, Rhipicephalus sanguineus. In each study, 6-8 male and 6-8 female purpose-bred, laboratory mongrels, terrier cross or Beagles were randomly assigned to one of two study groups (treated and untreated), based on pre-treatment parasite counts. Starting on the day before treatment, each dog was infested weekly with 50 ticks. Ticks were thumb counted at various time points after treatment and weekly infestations starting as early as 6h and continued at 12, 18 and 24h depending on the study. Ticks were removed and counted at 48 h after treatment and weekly infestations. CERTIFECT provided rapid and excellent control of pre-existing and newly acquired infestations of R. sanguineus with efficacy as high as 93% within the first 12h after a single topical treatment. Excellent control (>96%) of R. sanguineus as early as 18 h, following post treatment infestations was maintained for at least 35 days.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45323
Amitraz, PESTANAL®, analytical standard
C19H23N3