EMAIL THIS PAGE TO A FRIEND
Bioprocess and Biosystems Engineering
Assessment of two immobilized lipases activity and stability to low temperatures in organic solvents under ultrasound-assisted irradiation.
PMID 21779888