EMAIL THIS PAGE TO A FRIEND
NeuroImage
Extended characterisation of the serotonin 2A (5-HT2A) receptor-selective PET radiotracer 11C-MDL100907 in humans: quantitative analysis, test-retest reproducibility, and vulnerability to endogenous 5-HT tone.
PMID 21782029
Abstract
Scanning properties and analytic methodology of the 5-HT2A receptor-selective positron emission tomography (PET) tracer 11C-MDL100907 have been partially characterised in previous reports. We present an extended characterisation in healthy human subjects. 64 11C-MDL100907 PET scans with metabolite-corrected arterial input function were performed in 39 healthy adults (18-55 years). 12 subjects were scanned twice (duration 150 min) to provide data on plasma analysis, model order estimation, and stability and test-retest characteristics of outcome measures. All other scans were 90 min duration. 3 subjects completed scanning at baseline and following 5-HT2A receptor antagonist medication (risperidone or ciproheptadine) to provide definitive data on the suitability of the cerebellum as reference region. 10 subjects were scanned under reduced 5-HT and control conditions using rapid tryptophan depletion to investigate vulnerability to competition with endogenous 5-HT. 13 subjects were scanned as controls in clinical protocols. Pooled data were used to analyse the relationship between tracer injected mass and receptor occupancy, and age-related decline in 5-HT2A receptors. Optimum analytic method was a 2-tissue compartment model with arterial input function. However, basis function implementation of SRTM may be suitable for measuring between-group differences non-invasively and warrants further investigation. Scan duration of 90 min achieved stable outcome measures in all cortical regions except orbitofrontal which required 120 min. Binding potential (BPP and BPND) test-retest variability was very good (7-11%) in neocortical regions other than orbitofrontal, and moderately good (14-20%) in orbitofrontal cortex and medial temporal lobe. Saturation occupancy of 5-HT2A receptors by risperidone validates the use of the cerebellum as a region devoid of specific binding for the purposes of PET. We advocate a mass limit of 4.6 μg to remain below 5% receptor occupancy. 11C-MDL100907 specific binding is not vulnerable to competition with endogenous 5-HT in humans. Paradoxical decreases in BPND were found in right prefrontal cortex following reduced 5-HT, possibly representing receptor internalisation. Mean age-related decline in brain 5-HT2A receptors was 14.0±5.0% per decade, and higher in prefrontal regions. Our data confirm and extend support for 11C-MDL100907 as a PET tracer with very favourable properties for quantifying 5-HT2A receptors in the human brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3324
M100907, ≥98% (HPLC)
C22H28FNO3