EMAIL THIS PAGE TO A FRIEND
Toxicology
Evidence for aconitine-induced inhibition of delayed rectifier K(+) current in Jurkat T-lymphocytes.
PMID 21782880
Abstract
Aconitine (ACO) is a highly toxic diterpenoid alkaloid and known to exert the immunomodulatory action. However, whether it has any effects on ion currents in immune cells remains unknown. The effects of ACO and other related compounds on ion currents in Jurkat T-lymphocytes were investigated in this study. ACO suppressed the amplitude of delayed-rectifier K(+) current (I(K(DR))) in a time- and concentration-dependent manner. Margatoxin (100 nM), a specific blocker of K(V)1.3-encoded current, decreased the I(K(DR)) amplitude in these cells and the ACO-induced inhibition of I(K(DR)) was not reversed by 1-ethyl-2-benzimidazolinone (30 μM) or nicotine (10 μM). The IC(50) value for ACO-mediated inhibition of I(K(DR)) was 5.6 μM. ACO accelerated the inactivation of I(K(DR)) with no change in the activation rate of this current. Increasing the ACO concentration not only reduced the I(K(DR)) amplitude, but also accelerated the inactivation time course of the current. With the aid of minimal binding scheme, the inhibitory action of ACO on I(K(DR)) was estimated with a dissociation constant of 6.8 μM. ACO also shifted the inactivation curve of I(K(DR)) to a hyperpolarized potential with no change in the slope factor. Cumulative inactivation for I(K(DR)) was enhanced in the presence of ACO. In Jurkat cells incubated with amiloride (30 μM), the ACO-induced inhibition of I(K(DR)) remained unaltered. In RAW 264.7 murine macrophages, ACO did not modify the kinetics of I(K(DR)), although it suppressed I(K(DR)) amplitude. Taken together, these effects can significantly contribute to its action on functional activity of immune cells if similar results are found in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0034
1-EBIO, ≥98% (HPLC)
C9H10N2O
M8278 Margatoxin, ≥90% (HPLC)
C178H286N52O50S7