EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Biomarker response in the earthworm Lumbricus terrestris exposed to chemical pollutants.
PMID 21783228
Abstract
Earthworms are important organisms for the soil ecosystem. They are sensitive to toxic chemicals and represent useful bioindicator organisms for soil biomonitoring. Recently the use of biomarkers in earthworms has been increasingly investigated for soil monitoring and assessment purpose. The aim of the preset paper was to analyze the pollutant-induced response of a suite of cellular and biochemical biomarkers in the earthworm Lumbricus terrestris exposed to copper sulphate or methiocarb in OECD soil at the maximal concentrations recommended in agriculture. These responses were compared to lifecycle parameters such as survival, growth and reproduction. Granulocyte morphometric alteration, lysosomal membrane stability, metallothionein concentration, and acetylcholinesterase activity were considered. In either copper sulphate or methiocarb exposure conditions the mean percentage variation of the pollutant-induced molecular and cellular biomarkers was consistent with the whole organism end-point responses. In particular pollutant-induced granulocyte enlargement, detected in either copper sulphate or methiocarb exposed organisms, showed to be a potential general biomarker that may be directly linked to organism health. Compared to the other biological responses to pollutants, it showed high sensitivity to pollutant exposure suggesting its possible applications as a sensitive, simple, and quick general biomarker for monitoring and assessment applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36152
Methiocarb, PESTANAL®, analytical standard
C11H15NO2S