EMAIL THIS PAGE TO A FRIEND
Steroids
The steroid metabolome in lamotrigine-treated women with epilepsy.
PMID 21787799
Abstract
Epilepsy in women may be associated with reproductive disorders and alterations in serum steroid levels. Some steroids can be induced by epilepsy and/or treatment with antiepileptic drugs; however, there are still limited data available concerning this effect on the levels of other neuroactive steroid metabolites such as 3a-hydroxy-5a/b-reduced androstanes. To evaluate steroid alterations in women with epilepsy (WWE) on lamotrigine monotherapy. Eleven WWE and 11 age-matched healthy women underwent blood sampling in both phases of their menstrual cycles (MCs). The steroid metabolome, which included 30 unconjugated steroids, 17 steroid polar conjugates, gonadotropins, and sex hormone-binding globulin (SHBG), was measured using gas chromatography-mass spectrometry (GC-MS) and radioimmunoassay (RIA). WWE had lower cortisol levels (status p<0.001), but elevated levels of unconjugated 17-hydroxypregnenolone (status p<0.001). Progesterone was higher in the follicular menstrual phase (FP) in WWE than in the controls (status×menstrual phase p<0.05, Bonferroni multiple comparisons p<0.05), whereas 17-hydroxyprogesterone was higher in WWE in both menstrual phases (status p<0.001). The steroid conjugates were mostly elevated in WWE. The levels of 5α/β-reduced androstanes in WWE that were significantly higher than the controls were etiocholanolone (status p<0.001), 5α-androstane-3α,17β-diol (status p<0.001), and the 5α/β-reduced androstane polar conjugates (status p<0.001). WWE showed a trend toward higher circulating 3α-hydroxy-5α/β-reduced androstanes, increased activity of 17α-hydroxylase/17,20 lyase in the Δ(5)-steroid metabolic pathway, and increased levels of the steroid polar conjugates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H5002
17α-Hydroxypregnenolone
C21H32O3