EMAIL THIS PAGE TO A FRIEND
Langmuir : the ACS journal of surfaces and colloids
Gold nanoparticles grown on star-shaped block copolymer monolayers.
PMID 21790125
Abstract
We report on the growth of gold nanoparticles in polystyrene/poly(2-vinyl pyridine) (PS/P2VP) star-shaped block copolymer monolayers. These amphiphilic PS(n)P2VP(n) heteroarm star copolymers differ in molecular weight (149,000 and 529,000 Da) and the number of arms (9 and 28). Langmuir-Blodgett (LB) deposition was utilized to control the spatial arrangement of P2VP arms and their ability to reduce gold nanoparticles. The PS(n)P2VP(n) monolayer acted as a template for gold nanoparticle growth because of the monolayer's high micellar stability at the liquid-solid interface, uniform domain morphology, and ability to adsorb Au ions from the water subphase. UV-vis spectra and AFM and TEM images confirmed the formation of individual gold nanoparticles with an average size of 6 ± 1 nm in the P2VP-rich outer phase. This facile strategy is critical to the formation of ultrathin polymer-gold nanocomposite layers over large surface areas with confined, one-sided positioning of gold nanoparticles in an outer P2VP phase at polymer-silicon interfaces.