EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Occurrence and distribution of sulfonylurea and related herbicides in central Canadian surface waters 2006-2008.
PMID 21792538
Abstract
Surface water sampling in 2006-2008 measured the occurrence of sulfonylureas and related herbicides (SUs) during base flow conditions and wet weather events. Flumetsulam (29.2%), diuron (36.5%) and fomesafen (25.3%) were most frequently detected over the course of the study. Typical SU concentrations were in the low parts per trillion range; however, maximum concentrations of fomesafen (873 ng/L), linuron (856 ng/L) and diuron (2,900 ng/L) approached or exceeded 1 μg/L. The temporal trend in SUs showed a correlation with application periods. In general, detections of SUs were more frequent where rotation of row crops was more intense. Sampling during wet-weather events indicated potential for a range of SUs to be flushed into surface waters at relatively high concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46325
Fomesafen, PESTANAL®, analytical standard
C15H10ClF3N2O6S
36141
Linuron, PESTANAL®, analytical standard
C9H10Cl2N2O2