EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Titanocene(III) chloride mediated radical induced allylation of aldimines: formal synthesis of C-linked 4'-deoxy aza-disaccharide.
PMID 21793531
Abstract
Titanocene(III) chloride (Cp(2)TiCl) mediated radical induced allylation of aldimines for the preparation of homoallyl amines is described. The radical was generated from the allyl bromide using Cp(2)TiCl as the radical source. Formal synthesis of C(4)-C(5')-linked 4'-deoxy aza-disaccharide is demonstrated and a study toward the bicyclic skeleton of alkaloids was also accomplished. The radical initiator Cp(2)TiCl was prepared in situ from commercially available titanocene dichloride (Cp(2)TiCl(2)) and Zn dust in THF under argon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234826
Bis(cyclopentadienyl)titanium(IV) dichloride, 97%
C10H10Cl2Ti