EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Preparation of extracts from yeast.
PMID 2179664
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3148
2-Mercaptoethanol, for molecular biology, for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
C2H6OS
B6506
Benzamidine hydrochloride hydrate, ≥99%
C7H8N2 · ClH · xH2O
P5884 Brij® 58, average Mn ~1124
240850
D-Sorbitol, 99%
C6H14O6
S1876
D-Sorbitol, ≥98%
C6H14O6
85529
D-Sorbitol, BioUltra, ≥99.5% (HPLC)
C6H14O6
S6021
D-Sorbitol, for molecular biology, ≥98%
C6H14O6
S7547
D-Sorbitol, BioXtra, ≥98%
C6H14O6
S3889
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
C6H14O6
F9378 Ficoll® 400, Type 400-DL, lyophilized powder
F2637 Ficoll® 400, BioXtra, for molecular biology, lyophilized powder
F8016 Ficoll® 400, lyophilized powder, Type 400-DL, BioReagent, suitable for cell culture
F8636 Ficoll® 400, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
F1418 Ficoll® 400, BioXtra, Type 400-DL, lyophilized powder
F4375 Ficoll® PM 400, Type 400
L2884
Leupeptin, microbial, ≥90% (HPLC)
C20H38N6O4 · 0.5H2O4S
L8511
Leupeptin, synthetic, ≥85% (HPLC)
C20H38N6O4 · 0.5H2O4S
62070
Leupeptin, >4000 U/mg
C20H38N6O4 · 0.5H2O4S
L5793
Leupeptin, BioUltra, microbial, ≥95% (HPLC)
C20H38N6O4 · 0.5H2O4S
LEU-RO
Leupeptin, Ac-Leu-Leu-argininal x 1/2 H2SO4, synthetic
C20H38N6O4 · 0.5H2O4S
L9783
Leupeptin hydrochloride, microbial, ≥90% (HPLC)
C20H38N6O4 · HCl
L0649
Leupeptin hydrochloride, microbial, ≥70% (HPLC)
C20H38N6O4 · HCl
L2023
Leupeptin trifluoroacetate salt, ≥90% (HPLC), microbial
C20H38N6O4 · xC2HF3O2
L2524
Lyticase from Arthrobacter luteus, lyophilized powder, ≥2,000 units/mg protein, Protein ≥20 % by biuret
L4025
Lyticase from Arthrobacter luteus, lyophilized powder, ≥200 units/mg solid
SAE0098
Lyticase from Arthrobacter luteus, free of DNA contaminants, suitable for Microbiome research, ≥2000 units/mg protein, lyophilized powder
SRE0018
Lyticase from Arthrobacter Luteus, Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥2000 units/mg protein
P7626
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
C7H7FO2S
P7798 Polysucrose 400, powder