EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
Determination of active substances in binary mixture antiparasitic veterinary formulations by HPLC.
PMID 21796928
Abstract
The purpose of the study was to develop a simple, versatile HPLC method for the identification and quantification of praziquantel and ivermectin (in Equimax) or praziquantel and abamectin (in Abamitel Plus). A satisfactory separation was obtained using the Supelcosil LC-ABZ+ column in gradient system with a mobile phase A: acetonitrile / water in 40:60 ratio and phase B: acetonitrile. The UV detection was set at 245 nm. The correlation coefficient values (> or = 0,998) for all active substances confirmed that the calibration curves (peak area vs. concentration) are linear. The results of the quantification and the statistical evaluation confirmed that the method is accurate and precise. It can also be applied to confirm the identity of benzyl alcohol, methyl p-hydroxybenzoate and propyl p-hydroxybenzoate in Abamitel Plus formulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31732
Abamectin, PESTANAL®, analytical standard
M1650000
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C8H8O3
P5835
Propyl 4-hydroxybenzoate, BioXtra
C10H12O3
54789
Propyl 4-hydroxybenzoate, tested according to Ph.Eur.
C10H12O3
W295101
Propyl 4-hydroxybenzoate, ≥99%
C10H12O3
P3650000
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C10H12O3
PHR1010
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C10H12O3
59140C30 SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3 mm
59142C30 SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
59142C40 SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
59141 SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
59140-U SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
59142 SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
59170 SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
59611 SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
59610 SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
59545C30 SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm
59545-U SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
59544-U SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm