EMAIL THIS PAGE TO A FRIEND
Ecotoxicology and environmental safety
Differential physiological and biochemical responses of two cyanobacteria Nostoc muscorum and Phormidium foveolarum against oxyfluorfen and UV-B radiation.
PMID 21798597
Abstract
In the present study, degree of tolerance and tolerance strategies of two paddy field cyanobacteria viz. Nostoc muscorum and Phormidium foveolarum against oxyfluorfen (10 and 20 μg ml(-1)) and UV-B (7.2 kJ m(-2)d(-1)) stress were investigated. Oxyfluorfen and UV-B decreased growth, photosynthesis, nutrient uptake, nitrate reductase, acid and alkaline phosphatase activities, which accompanied with the increase in the level of oxidative stress. However, growth was more affected in N. muscorum than P. foveolarum. Antioxidants exhibited differential responses against oxyfluorfen and UV-B stress. Ascorbate and proline levels were higher in P. foveolarum. A protein of 66 kDa was expressed in N. muscorum, however, it was absent in P. foveolarum than those of N. muscorum. Besides this, a protein of 29 kDa appeared in P. foveolarum under all the treatments, but it was present only in control cells of N. muscorum cells. Overall results indicated resistant nature of P. foveolarum against oxyfluorfen and UV-B stress in comparison to N. muscorum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35031
Oxyfluorfen, PESTANAL®, analytical standard
C15H11ClF3NO4