EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
Fluoroquinolone-resistant tuberculosis at a medical centre in Taiwan, 2005-10.
PMID 21798881
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E6005
Ethionamide
C8H10N2S
09415
Sodium 4-aminosalicylate dihydrate, ≥97.0% (T)
C7H6NNaO3 · 2H2O