EMAIL THIS PAGE TO A FRIEND
Nature protocols
Continuous-flow reactor-based synthesis of carbohydrate and dihydrolipoic acid-capped quantum dots.
PMID 21799489
Abstract
A detailed protocol for the large-scale synthesis of carbohydrate and dihydrolipoic acid (DHLA)-coated CdSe/ZnS and CdTe/ZnS nanoparticles using continuous flow reactors is described here. Three continuous flow microreaction systems, operating at three different temperatures, are used for the synthesis of mannose-, galactose- or DHLA-functionalized quantum dots (QDs). In the first step of synthesis, the CdSe and CdTe nanoparticles are prepared. The size and spectral properties of the CdSe core of the nanoparticles are controlled by adjustment of the residence time and the temperature. As a second step, the zinc sulfide capping under homogenous conditions is carried out at a substantially lower temperature than is required for nanoparticle growth in batch processes. Finally, the trioctylphosphine/oleic acid ligand is effectively replaced with either carbohydrate PEG-thiol moieties or DHLA at 60 °C. This new protocol allows the synthesis of biologically active fluorescent QDs in 4 d.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8260
Lipoic acid, reduced, analytical standard
C8H16O2S2