EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
One-pot synthesis of an organic-inorganic hybrid affinity monolithic column for specific capture of glycoproteins.
PMID 21799960
Abstract
An inorganic-organic hybrid affinity monolithic column was synthesized by a novel "one-pot" approach. The resulting hybrid affinity monoliths have potential applications in specific recognition and enrichment of glycoproteins.