EMAIL THIS PAGE TO A FRIEND
Pest management science
Absorption, translocation and metabolism of metamitron in Chenopodium album.
PMID 21800410
Abstract
In recent years, common lambsquarters (Chenopodium album L.) populations from sugar beet fields in different European countries have responded as resistant to the as-triazinone metamitron. The populations have been found to have the same D1 point mutation as known for atrazine-resistant biotypes (Ser264 to Gly). However, pot experiments revealed that metamitron resistance is not as clear-cut as observed with triazine resistance in the past. The objectives of this study were to clarify the absorption, translocation and metabolic fate of metamitron in C. album. Root absorption and foliar absorption experiments showed minor differences in absorption, translocation and metabolism of metamitron between the susceptible and resistant C. album populations. A rapid metabolism in the C. album populations was observed when metamitron was absorbed by the roots. The primary products of metamitron metabolism were identified as deamino-metamitron and metamitron-N-glucoside. PABA, known to inhibit the deamination of metribuzin, did not alter the metabolism of metamitron, and nor did the cytochrome P450 inhibitor PBO. However, inhibition of metamitron metabolism in the presence of the cytochrome P450 inhibitor ABT was demonstrated. Metamitron metabolism in C. album may act as a basic tolerance mechanism, which can be important in circumstances favouring this degradation pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3940
1-Aminobenzotriazole
C6H6N4
36154
Metamitron, PESTANAL®, analytical standard
C10H10N4O