EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of the scalarane sesterterpenoid 16-deacetoxy-12-epi-scalarafuranacetate.
PMID 21800844
Abstract
The marine natural product 16-deacetoxy-12-epi-scalarafuranacetate, isolated from Spongua officinalis , was synthesized in 18 linear steps, starting from (-)-sclareol, with high stereoselectivity and an overall yield of 6.1%. The intermediate 16-deacetoxy-12-epi-scalarafuran could be easily transformed into a series of natural scalarane sesterterpenoids in a few steps.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357995
Sclareol, 98%
C20H36O2