EMAIL THIS PAGE TO A FRIEND
Trends in pharmacological sciences
How reserpine and chlorpromazine act: the impact of key discoveries on the history of psychopharmacology.
PMID 2180160
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0200000
Reserpine, European Pharmacopoeia (EP) Reference Standard
C33H40N2O9