EMAIL THIS PAGE TO A FRIEND
The International Journal of Biochemistry & Cell Biology
Nonsense mutations of the bHLH transcription factor TWIST2 found in Setleis Syndrome patients cause dysregulation of periostin.
PMID 21801849