EMAIL THIS PAGE TO A FRIEND
Respiratory physiology & neurobiology
Congenital central hypoventilation syndrome and desogestrel: a call for caution: addendum to "C. Straus, H. Trang, M.H. Becquemin, P. Touraine, T. Similowski, Chemosensitivity recovery in Ondine's curse syndrome under treatment with desogestrel" [Respir. Physiol. Neurobiol. 171 (2010) 171-174].
PMID 21801857
Abstract
Patients suffering from congenital central hypoventilation syndrome (CCHS) depend on mechanical ventilation during sleep, from birth and throughout life. They lack CO₂-chemosensitivity. Hope has recently been raised by serendipitous observations of chemosensitivity recovery under treatment by desogestrel, a very potent progestin (Straus et al., 2010). Caution is however needed, because this effect could depend on dose, idiosyncrasies, or be transient. Desogestrel should not be prescribed to CCHS patients with a respiratory purpose until the results of a pending clinical trial (clinicaltrials.gov. NCT01243697) are available.