EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Synthesis and evaluation of novel tetrapropoxycalix[4]arene enones and cinnamates for protection from ultraviolet radiation.
PMID 21803597
Abstract
A series of novel calix[4]arene enones (5-7) and cinnamates (12-14) have been synthesized and evaluated for ensuring protection from ultraviolet radiation (UVR). Spectroscopic analyses has revealed that compound 6 absorbs ultraviolet radiations between 280 and 350 nm with an absorption maximum at 312 nm. Its molar absorption coefficient (ε) (>5 × 10(4)M(-1)cm(-1)) and bandwidth are larger than those for the commercially used sun protectants (oxybenzone (OB), 2-ethylhexyl 4-methoxycinnamate (OMC) and avobenzone). The in vitro Sun Protection Factor (SPF) measurement revealed an SPF of 5.2 at 2% concentration of 6 in home made emulsion formulations while combination of 2% each of 6 and OMC gave an SPF of 8.8. Lower sun protection seems to be compensated by significant protection from more harmful UVA radiations (UVA/UVB absorbance ratio of 0.62).
Related Materials