EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Synthesis, characterization, crystal structure and DFT studies on 1',3'-dihydrospiro[fluorene-9,2'-perimidine].
PMID 21803646
Abstract
The title compound, 1',3'-dihydrospiro[fluorene-9,2'-perimidine] has been synthesized and characterized by NMR, ESI-MS, IR, elemental analysis, UV-vis and fluorescence spectroscopy. The crystal structures of the title compound and its co-crystal with 9-fluorenone have also been determined by X-ray single crystal diffraction. Density functional theory (DFT) calculations and vibrational frequencies have been performed at B3LYP/6-31G* level. The comparisons between the experimental vibrational frequencies and the predicted data show that B3LYP/6-31G* method can simulate the IR of the title compound on the whole. The theoretical electronic absorption spectra have been calculated by using TD-DFT method and compared with the experimental result. The solid-fluorescence determination of the title compound reveals two emission bans at 430 and 590 nm while its co-crystal reveals only one emission band at 590 nm.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F1506
9-Fluorenone, 98%
C13H8O