EMAIL THIS PAGE TO A FRIEND
Organic letters
A near-infrared fluorescence turn-on sensor for sulfide anions.
PMID 21809838
Abstract
The first NIR fluorescent sensor for sulfide anions was constructed based on the displacement approach. The sensing ensemble is composed of a cyanine dye, a piperazine linker, an 8-aminoquinoline ligand, and copper. The favorable attributes of the sensor include a large NIR fluorescence turn-on signal in aqueous ethanol, high sensitivity, and high selectivity. The transition-metal-based displacement strategy may open an avenue for development of NIR fluorescent sensors for a wide variety of anion targets.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
260789
8-Aminoquinoline, 98%
C9H8N2