EMAIL THIS PAGE TO A FRIEND
Journal of physics. Condensed matter : an Institute of Physics journal
Evidence of two distinct dynamical freezing processes in single-layered perovskite La(0.7)Sr(1.3)CoO4.
PMID 21811008
Abstract
The static and dynamic features of magnetization have been investigated in terms of dc magnetization, ac susceptibility, and memory effects on single-layered perovskite La(0.7)Sr(1.3)CoO(4). The results indicate that short-range ferromagnetic clusters coexist with the glassy magnetic state, i.e., a cluster-glass phase between T(f) and T(G) and a spin-glass-like phase below T(f). The clear evidence of memory effects in the dc magnetization is also observed below T(f) and T(G), respectively. These two glassy states with cooperative relaxation processes may derive from the different interaction processes among nanoscale ferromagnetic clusters mediated by the matrix, which is influenced by changing the spin state of Co(3+) ions in spontaneously phase-separated cobaltite.