EMAIL THIS PAGE TO A FRIEND
The American journal of psychiatry
The NIMH-CATIE Schizophrenia Study: what did we learn?
PMID 21813492
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6402
Perphenazine
C21H26ClN3OS