EMAIL THIS PAGE TO A FRIEND
Journal of virology
Identification of chemoattractive factors involved in the migration of bone marrow-derived mesenchymal stem cells to brain lesions caused by prions.
PMID 21813601