EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Phenylacetonitrile from the giant knotweed, Fallopia sachalinensis, infested by the Japanese beetle, Popillia japonica, is induced by exogenous methyl jasmonate.
PMID 21814160
Abstract
Phenylacetonitrile, (E)-β-ocimene, linalool, (E)-4,8-dimethyl-1,3,7-nonatriene and (E,E)-α-farnesene were identified as Japanese beetle, Popillia japonica, feeding-induced volatiles from the leaves of the giant knotweed, Fallopia sachalinensis, but not by mechanical damage. Volatile emission was also induced by treatment with a cellular signaling molecule, methyl jasmonate. These results suggest that volatiles will be synthesized de novo by a biotic elicitor from P. japonica oral secretion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B19401
Benzyl cyanide, 98%
C8H7N
W353901
Ocimene, mixture of isomers, stabilized, ≥90%
C10H16