EMAIL THIS PAGE TO A FRIEND
Pharmacology & therapeutics
The metabolic basis of 3-methylindole-induced pneumotoxicity.
PMID 2181487
Abstract
3-Methylindole (3MI), an abnormal metabolite of tryptophan, causes acute pulmonary edema and emphysema. 3MI toxicity is species-, tissue- and cell-specific and is an excellent model for understanding the processes of chemically-induced lung injury. Experimental evidence showed that 3MI is metabolically activated by both microsomal cytochrome P-450-dependent mixed function oxidase (MFO) and prostaglandin H synthase (PHS) systems in the lung. Formation of a free radical intermediate during 3MI metabolism is the initial chemical event which is responsible for the pneumotoxicity. 3MI free radicals bind covalently to microsomal protein and induce lipid peroxidation. Microsomal enzymes which regulate the glycogen and phospholipid biosynthesis in the lung are altered during the cellular repair processes after 3MI-induced lung injury. Inhibition of cellular differentiation from Type II to Type I cells and impaired surfactant function may be crucial to the disease process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M51458
3-Methylindole, 98%
C9H9N
W301912
Skatole, ≥98%
C9H9N