EMAIL THIS PAGE TO A FRIEND
Journal of pediatric orthopedics. Part B
Iliac osteomyelitis in a newborn: a case report.
PMID 21817921
Abstract
Pelvis is a rare location of the hematogeneous osteomyelitis during the newborn period. Successful cure of osteomyelitis during the newborn period is dependent on a fast and true diagnosis and on sufficient treatment. In this case report, iliac osteomyelitis of a newborn that was clinically suspected on the fifth day, was diagnosed radiologically and was found to be caused by methicillin-sensitive Staphylococcus aureus, is presented. According to our knowledge, this is the youngest osteomyelitis case in the literature and the third presented iliac osteomyelitis case of the newborn period.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4237
Cefuroxime Axetil
C20H22N4O10S