EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Isoconiferoside, a new phenolic glucoside from seeds of Panax ginseng.
PMID 21818060
Abstract
A new phenolic glucoside, isoconiferoside (1), was isolated from the seeds of Panax ginseng (Araliaceae). The structure was determined to be 9-O-[β-D-glucopyranosyl-(1 --> 6)-β-D-glucopyranosyl]-trans-coniferyl alcohol based on spectroscopic analyses (1H- and 13C-NMR, DEPT, COSY, HMQC, and HMBC) and acid hydrolysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223735
Coniferyl alcohol, 98%
C10H12O3