EMAIL THIS PAGE TO A FRIEND
Experimental and clinical transplantation : official journal of the Middle East Society for Organ Transplantation
Flupirtine-induced hepatic failure requiring orthotopic liver transplant.
PMID 21819373
Abstract
We present the case of a 48-year-old otherwise healthy man who required an urgent liver transplant owing to acute liver failure after flupirtine treatment. After 3 months of daily flupirtine intake as treatment for pseudoradicular pain syndrome, he presented at our institution with signs of jaundice and hepatic encephalopathy. Laboratory results showed elevated liver transaminases, and the liver histopathology supported the assumed drug-induced liver injury. After listing him for an urgent liver transplant, he was given a liver graft from a 21-year-old man. Despite a rejection episode on day 11 after the surgery (which was successfully treated by steroid pulse therapy), the postoperative course was uneventful and the patient recovered completely. To the best of our knowledge, this is the first report of a liver transplant for acute liver failure after taking flupirtine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8927
Flupirtine maleate salt, ≥98% (HPLC)
C15H17FN4O2 · C4H4O4