EMAIL THIS PAGE TO A FRIEND
Thrombosis research
Tamibarotene-induced low-grade reversible intravascular coagulation in a patient with acute promyelocytic leukemia.
PMID 21820159
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3205
Tamibarotene, ≥98% (HPLC)
C22H25NO3