EMAIL THIS PAGE TO A FRIEND
Journal of pediatric urology
Intermediate to long-term follow-up indicates low risk of recurrence after Double HIT endoscopic treatment for primary vesico-ureteral reflux.
PMID 21820358
Abstract
Follow-up of patients undergoing dextranomer/hyaluronic acid injection for vesico-ureteral reflux (VUR) is controversial. The purpose of our study was to test the hypothesis that patients undergoing the double hydrodistention-implantation technique (Double HIT) have a higher clinical and radiographic success rate. Patients undergoing Double HIT endoscopic injection for VUR were prospectively identified. Patients underwent an ultrasound at 6 weeks to assess the implants, and, if visible, prophylactic antibiotics were discontinued and patients were scheduled for a 1-year voiding cystourethrogram (VCUG). Radiographic success was defined as a negative VCUG and clinical success as no febrile urinary tract infections at 1 year. A total of 54 patients underwent endoscopic injection for VUR. Twenty-five (51%) were compliant with the 1 year follow-up; 18 non-compliant patients were contacted and their clinical status assessed. Thirty patients eventually completed the 1-year VCUG at a mean of 12.2 months (range 10-20). Among the 60% of patients with 1-year radiographic follow-up, 2 had persistent VUR for a radiologic success rate of 93%. All radiographic failures were infection-free. Of the 80% (43/54) of patients with available clinical data, 3 (7%) had afebrile UTI for a clinical success rate of 93%. The Double HIT leads to a 93% clinical and 93% radiographic intermediate/long-term success rate. With this technique, better outcomes were achieved with fewer recurrences than previously reported. These favorable results challenge the need for postoperative VCUG in asymptomatic patients after the Double HIT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000744 Dextranomer, European Pharmacopoeia (EP) Reference Standard