EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
Hippocampal AMPA autoreceptors positively coupled to NMDA autoreceptors traffic in a constitutive manner and undergo adaptative changes following enriched environment training.
PMID 21820454
Abstract
α-Amino-3-hydroxy-5-methyl-4-isoxazole propionate (AMPA) autoreceptors exist on glutamate hippocampal terminals. Aimed at investigating whether these autoreceptors traffic constitutively, (S)AMPA-evoked [(3)H]D-ASP release from synaptosomes enriched with peptides that impede the interaction of GluA2 subunits with cytosolic proteins involved in receptor movements [namely Glutamate Receptor-Interacting Protein (GRIP), Protein Interacting with C kinase 1 (PICK1), N-ethyl-maleimide-Sensitive Fusion protein NSF proteins] was monitored. (S)AMPA alone had no effect on the spontaneous release of [(3)H]D-ASP from control synaptosomes, but became efficacious in the presence of cyclothiazide or when preventing GluA2/GRIP/PICK1, but not GluA2/NSF, interaction. Hippocampal glutamatergic terminals also possess NMDA autoreceptors. 10 μM NMDA/1 μM glycine-induced [(3)H]D-ASP release was concentration-dependently increased by (S)AMPA. Cyclothiazide potentiated the 10 μM NMDA/1 μM glycine/50 μM (S)AMPA-induced [(3)H]D-ASP overflow, while NBQX halved and MK-801 abolished it, suggesting NMDA-AMPA autoreceptor cross-talk. Western Blot analysis of sub-synaptic fractions confirmed presynaptic GluN2B-GluA2/3 co-localization. Impeding GluA2/GRIP/PICK1 interaction facilitated the NMDA/glycine/(S)AMPA-induced release of [(3)H]D-ASP, while competing for GluA2/NSF interaction reduced it, indicating that NMDA receptor favours AMPA receptor insertion in synaptosomal plasmamembranes. Finally, rearing mice in enriched environment unveiled the (S)AMPA-induced release of [(3)H]D-ASP, but leaved unmodified that caused by NMDA/glycine. The NBQX-sensitive, 50 μM (S)AMPA-evoked release of [(3)H]D-ASP was insensitive to cyclothiazide and to peptide interfering with GluA2/GRIP/PICK1 interaction but was addictive to that caused by NMDA/glycine. Presynaptic GluA2/3 immunoreactivity in EE hippocampal terminals was increased, while GluN2B was unchanged. We conclude that hippocampal AMPA autoreceptors positively coupled to NMDA autoreceptors traffic in a constitutive manner and undergo functional up-regulation in EE animals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9847
Cyclothiazide
C14H16ClN3O4S2
219096
D-Aspartic acid, ReagentPlus®, 99%
C4H7NO4