EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Cork-based activated carbons as supported adsorbent materials for trace level analysis of ibuprofen and clofibric acid in environmental and biological matrices.
PMID 21820664
Abstract
In this contribution, powdered activated carbons (ACs) from cork waste were supported for bar adsorptive micro-extraction (BAμE), as novel adsorbent phases for the analysis of polar compounds. By combining this approach with liquid desorption followed by high performance liquid chromatography with diode array detection (BAμE(AC)-LD/HPLC-DAD), good analytical performance was achieved using clofibric acid (CLOF) and ibuprofen (IBU) model compounds in environmental and biological matrices. Assays performed on 30 mL water samples spiked at the 25.0 μg L(-1) level yielded recoveries around 80% for CLOF and 95% for IBU, under optimized experimental conditions. The ACs textural and surface chemistry properties were correlated with the results obtained. The analytical performance showed good precision (<15%), suitable detection limits (0.24 and 0.78 μg L(-1) for CLOF and IBU, respectively) and good linear dynamic ranges (r(2)>0.9922) from 1.0 to 600.0 μg L(-1). By using the standard addition methodology, the application of the present approach to environmental water and urine matrices allowed remarkable performance at the trace level. The proposed methodology proved to be a viable alternative for acidic pharmaceuticals analysis, showing to be easy to implement, reliable, sensitive and requiring low sample volume to monitor these priority compounds in environmental and biological matrices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
197777
2-(p-Chlorophenoxy)-2-methylpropionic acid, 97%
C10H11ClO3