EMAIL THIS PAGE TO A FRIEND
Archivos de la Sociedad Espanola de Oftalmologia
[Sterile non-traumatic corneal perforation treated with Tachosil®].
PMID 21821194
Abstract
An 83 year-old male reported to the emergency room with red eye and ocular pain. The slit-lamp examination showed a central corneal perforation of 1mm in diameter, with no associated infection or inflammatory process. The patient had no history of trauma. It was decided to apply a patch of Tachosil(®) at the site of corneal perforation. After 36 hours of occlusion, a layer of fibrin covered the perforation, closing the defect. Successful healing of the corneal perforation was observed in the follow-up, with no recurrences of the process. Tachosil(®) is collagen sponge with human fibrin and human thrombin used in cardiological and neurosurgical surgery for haemostasis and the occlusion of vessels and meninges. We present the first indication in ophthalmology of Tachosil(®) as an effective alternative in the treatment of sterile non-traumatic corneal perforations less than 1.5 mm in size.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6013
Medroxyprogesterone
C22H32O3
46411
Medroxyprogesterone, VETRANAL, analytical standard
C22H32O3