EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section F, Structural biology and crystallization communications
Expression, purification, crystallization and preliminary crystallographic analysis of Cg1458: a novel oxaloacetate decarboxylase from Corynebacterium glutamicum.
PMID 21821907
Abstract
Oxaloacetate decarboxylase catalyses the decarboxylation of oxaloacetate to pyruvate and CO(2). Recently, the Corynebacterium glutamicum gene product Cg1458 was determined to be a soluble oxaloacetate decarboxylase. To elucidate the mechanism of oxaloacetate decarboxylation by Cg1458, recombinant Cg1458 was purified and crystallized. The best crystal was grown from 0.2 M MgCl(2), 0.1 M Bis-Tris pH 6.0, 25%(w/v) polyethylene glycol 3350 using the hanging-drop method. The crystals belonged to space group P4(3)2(1)2, with unit-cell parameters a = b = 124.1, c = 73.6 Å. The crystals are most likely to contain a dimer in the asymmetric unit, with a V(M) value of 2.27 Å(3) Da(-1). A full data set was collected at 1.9 Å resolution using synchrotron radiation on beamline BL17U of SSRF, Shanghai, China. Structure-solution attempts by molecular replacement were successful with PDB entries 3qdf or 2dfu as the template.
Related Materials