EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Captafol.
PMID 21822319
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45365
Captafol, PESTANAL®, analytical standard
C10H9Cl4NO2S
32054
Captan, PESTANAL®, analytical standard
C9H8Cl3NO2S