EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
Cobalt-tungsten carbide: powders and hard metals.
PMID 21822320
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241881
Tungsten(IV) carbide, powder, 2 μm, ≥99%
CW