EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on the diterpenoids of Wedelia prostrata].
PMID 21823454
Abstract
To study the chemical constituents of Wedelia prostrata. Chromatography and spectroscopic analysis were employed to isolate and elucidate the chemical constituents in the plant. Seven compounds were obtained and identified as ent-kaur-16-en-19-oic acid (I), ent-kaur-16alpha, 17-dihydroxy-19-oic acid (II), ent-kaur-15alpha-senecioyloxy-16-en-19-oic acid (III), beta-sitosterol (IV), stigmasterol (V), stigmasterol-3-O-beta-D-glucopyranoside (VI) and n-hexacosanol (VII). Compounds I-III are isolated from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2139
1-Hexacosanol, ≥97% (capillary GC)
C26H54O