EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on the chemical constituents from stems and leaves of Lonicera macranthoides].
PMID 21823476
Abstract
To research the chemical constituents from stems and leaves of Lonicera macranthoides. Various column chromatographies were employed to isolate and purify the constituents. Their structures were elucidated by spectral analysis (IR, MS, 1H-NMR, 13 C-NMR) and chemical evidence. Nine constituents were obtained and identified as loganin (I), loganic acid (II), morroniside (III),7-O-ethyl-morroniside (IV), scopoletin (V), caffeic acid (VI), chlorogenic acid (VII), beta-sitosterol (VIII), daucosterol (IX). Compounds I-VI are isolated from the plant for the first time. All the compounds are found for the first time from the stems and leaves of Lonicera macranthoides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00231
Loganic acid, ≥95% (LC/MS-ELSD)
C16H24O10
36483
Loganin, analytical standard
C17H26O10