EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on the chemical constituents of Salsola collina].
PMID 21823480
Abstract
To study the chemical constituents of Salsola collina. The compounds were isolated by column chromatography, and their structures were identified by physical and chemical properties and spectral data. Seven compounds were isolated and identified as n-lignoceric acid (1), n-dotriacontanic acid (2), beta-sitosterol (3), 5, 2'-dihydroxy-6, 7-methylenedioxy-isoflavone (4), vanillic acid (5), (-)syringaresinol 4,4'-bis-O-beta-D-glucopyranoside (6), salicylic acid (7). The compounds of 1, 2, 4, 5 and 6 are isolated from the plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6641
Lignoceric acid, ≥99% (GC)
C24H48O2