EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on the chemical constituents of Artemisia lavandulaefolia].
PMID 21823482
Abstract
To investigate the chemical constituents from aerial part of Aremisia lavandulaefolia. Compounds were isolated by using silicagel, Sephadex LH-20 chromatography and their structures were elucidated by means of spectral data analysiy. Eight compounds were isolated and identifies as P-amyrin(1), P-sitosterol(2), ursolic acid(3), erpatilin(4), naringenin(5), apigenin(6), luteolin(7), quercetin(8). All these compounds are isolated from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53017
α-Amyrin, analytical standard
C30H50O
09236
β-Amyrin, analytical standard
C30H50O