EMAIL THIS PAGE TO A FRIEND
Liver international : official journal of the International Association for the Study of the Liver
New insights into the management of hepato-renal syndrome.
PMID 21824281
Abstract
Hepato-renal syndrome (HRS) is a serious complication in patients with advanced liver disease indicating a very poor prognosis. Nevertheless, effective treatment strategies have been introduced into clinical practice over the last decade. The combined treatment with terlipressin/albumin has been proven to be effective in most published studies and should be regarded as the standard of care. Norepinephrine or midodrine are possible alternatives when terlipressin is not available. Although there is presently not enough evidence to recommend extracorporeal liver support therapy as a standard procedure in patients with HRS, these concepts constitute promising options that need to be studied in prospective clinical trials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8277
Midodrine hydrochloride, analytical standard, for drug analysis
C12H18N2O4 · HCl