EMAIL THIS PAGE TO A FRIEND
Report on carcinogens : carcinogen profiles
1-Amino-2-methylanthraquinone.
PMID 21829246
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42994
Disperse Orange 11, analytical standard
C15H11NO2