EMAIL THIS PAGE TO A FRIEND
Amino acids
The reaction of methionine with hydroxyl radical: reactive intermediates and methanethiol production.
PMID 21830117
Abstract
The mechanisms of reaction of methionine with hydroxyl radical are not fully understood. Here, we unequivocally show using electron paramagnetic resonance spin-trapping spectroscopy and GC-FID and GC-MS, the presence of specific carbon-, nitrogen- and sulfur-centered radicals as intermediates of this reaction, as well as the liberation of methanethiol as a gaseous end product. Taking into account the many roles that methionine has in eco- and biosystems, our results may elucidate redox chemistry of this amino acid and processes that methionine is involved in.