EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Antioxidant and urease inhibitory C-glycosylflavonoids from Celtis africana.
PMID 21830883
Abstract
Two new C-glycosylflavonoids celtisides A (1) and B (2) have been isolated from n-butanol-soluble fraction of Celtis africana, along with five known C-glycosylflavonoids vitexin (3), orientin (4), isoswertiajaponin (5), isoswertisin (6), and 2″-O-rhamnosyl vitexin (7) reported for the first time from this species. Their structures were assigned from 1D and 2D NMR spectra. These compounds were investigated for biological activities and showed significant antioxidant and urease inhibitory activities.
Related Materials